3-Thiophenemethanol, 97%
3-Thiophenemethanol, 97%
3-Thiophenemethanol, 97%
Thermo Scientific Chemicals

3-Thiophenemethanol, 97%

CAS: 71637-34-8 | C5H6OS | 114.16 g/mol
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 442240050
Price (JPY)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS71637-34-8
IUPAC Name(thiophen-3-yl)methanol
Molecular FormulaC5H6OS
InChI KeyBOWIFWCBNWWZOG-UHFFFAOYSA-N
SMILESOCC1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only