D-Phenylalanine methyl ester hydrochloride, 98%
D-Phenylalanine methyl ester hydrochloride, 98%
D-Phenylalanine methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

D-Phenylalanine methyl ester hydrochloride, 98%

CAS: 13033-84-6 | C10H14ClNO2 | 215.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 442620050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS13033-84-6
IUPAC Namemethyl (2R)-2-amino-3-phenylpropanoate hydrochloride
Molecular FormulaC10H14ClNO2
InChI KeySWVMLNPDTIFDDY-SBSPUUFOSA-N
SMILESCl.COC(=O)[C@H](N)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Specific optical rotation-32° to -38° (20°C, 589 nm) (c=1 in EtOH)
Appearance (Form)Powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only