3-Amino-5-bromobenzotrifluoride, 97%
3-Amino-5-bromobenzotrifluoride, 97%
3-Amino-5-bromobenzotrifluoride, 97%
Thermo Scientific Chemicals

3-Amino-5-bromobenzotrifluoride, 97%

CAS: 54962-75-3 | C7H5BrF3N | 240.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 442660050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS54962-75-3
IUPAC Name3-bromo-5-(trifluoromethyl)aniline
Molecular FormulaC7H5BrF3N
InChI KeyHJTLKVYOWNTDPF-UHFFFAOYSA-N
SMILESNC1=CC(Br)=CC(=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Colorless to brown
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only