OpenArray™ Plate Adaptor for Dual Flat Block GeneAmp™ PCR System 9700
The OpenArray™ Plate Adaptor for Dual Flat Block GeneAmp™ PCR System 9700 serves to secure up to eight TaqMan™ OpenArray™Read more
Have Questions?
Catalog Number
Quantity
4426824
1 adaptor
Catalog number 4426824
Price (USD)
492.00
Each
Quantity:
1 adaptor
Price (USD)
492.00
Each
The OpenArray™ Plate Adaptor for Dual Flat Block GeneAmp™ PCR System 9700 serves to secure up to eight TaqMan™ OpenArray™ Genotyping Plates in place for PCR amplification. Use the Plate Adaptor with the Dual Flat Block GeneAmp™ PCR System 9700 System and OpenArray™ Plate Compression Pad for Dual Flat Block GeneAmp™ PCR System 9700.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)PCR
For Use With (Equipment)GeneAmp 9700
Plate CompatibilityOpenArray™ Plates
Product LineGeneAmp, OpenArray
Quantity1 adaptor
Shipping ConditionRoom Temperature
Product TypeThermal Cycler Accessories
Unit SizeEach
Contents & Storage
Required adaptor for using OpenArray™ plate holder with the GeneAmp 9700 Dual flat block PCR system
Frequently asked questions (FAQs)
I have an Applied Biosystems GeneAmp PCR System 9700. What additional accessories do I need to perform off-line cycling for Applied Biosystems QuantStudio OpenArray plates?
You need an Applied Biosystems OpenArray Plate Adaptor (Part Number 4426824). You do not need a compression pad.