1-Bromo-4-ethynylbenzene, 98%
1-Bromo-4-ethynylbenzene, 98%
1-Bromo-4-ethynylbenzene, 98%
Thermo Scientific Chemicals

1-Bromo-4-ethynylbenzene, 98%

CAS: 766-96-1 | C8H5Br | 181.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4427900101 gGlass bottle
Catalog number 442790010
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Chemical Identifiers
CAS766-96-1
IUPAC Name1-bromo-4-ethynylbenzene
Molecular FormulaC8H5Br
InChI KeyLTLVZQZDXQWLHU-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or solid
Melting point62°C to 68°C
Appearance (Color)White to yellow
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only