Ethylene glycol dimethyl ether, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, stabilized
Ethylene glycol dimethyl ether, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, stabilized
Ethylene glycol dimethyl ether, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, stabilized
Thermo Scientific Chemicals

Ethylene glycol dimethyl ether, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, stabilized

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 442980010
Price (USD)
292.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
292.00
Each
Chemical Identifiers
CAS110-71-4
IUPAC Name1,2-dimethoxyethane
Molecular FormulaC4H10O2
InChI KeyXTHFKEDIFFGKHM-UHFFFAOYSA-N
SMILESCOCCOC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Water=<0.005 %
Appearance (Form)Liquid
GC>=99.4 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only