1,2-Dichloroethane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
1,2-Dichloroethane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
1,2-Dichloroethane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™
Thermo Scientific Chemicals

1,2-Dichloroethane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™

CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 443010010
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS107-06-2
IUPAC Name1,2-dichloroethane
Molecular FormulaC2H4Cl2
InChI KeyWSLDOOZREJYCGB-UHFFFAOYSA-N
SMILESClCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.4 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.005 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only