Methyl pyrazine-2-carboxylate, 97%
Methyl pyrazine-2-carboxylate, 97%
Methyl pyrazine-2-carboxylate, 97%
Thermo Scientific Chemicals

Methyl pyrazine-2-carboxylate, 97%

CAS: 6164-79-0 | C6H6N2O2 | 138.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 443170010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS6164-79-0
IUPAC Namemethyl pyrazine-2-carboxylate
Molecular FormulaC6H6N2O2
InChI KeyTWIIRMSFZNYMQE-UHFFFAOYSA-N
SMILESCOC(=O)C1=CN=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light orange
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point57°C to 63°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only