(R)-(+)-4-Hydroxy-2-pyrrolidinone, 97%
(R)-(+)-4-Hydroxy-2-pyrrolidinone, 97%
(R)-(+)-4-Hydroxy-2-pyrrolidinone, 97%
Thermo Scientific Chemicals

(R)-(+)-4-Hydroxy-2-pyrrolidinone, 97%

CAS: 22677-21-0 | C4H7NO2 | 101.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 443820050
Price (USD)
434.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
434.00
Each
Chemical Identifiers
CAS22677-21-0
IUPAC Name(4R)-4-hydroxypyrrolidin-2-one
Molecular FormulaC4H7NO2
InChI KeyIOGISYQVOGVIEU-GSVOUGTGSA-N
SMILESO[C@H]1CNC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=96.0 %
Appearance (Color)White to off-white
Specific optical rotation+54° to +60° (20°C, 589 nm) (c=0.36 in water)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only