2-(Dimethylamino)thiazole-5-carboxaldehyde, 97%
2-(Dimethylamino)thiazole-5-carboxaldehyde, 97%
2-(Dimethylamino)thiazole-5-carboxaldehyde, 97%
Thermo Scientific Chemicals

2-(Dimethylamino)thiazole-5-carboxaldehyde, 97%

CAS: 1005-28-3 | C6H8N2OS | 156.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 444040010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1005-28-3
IUPAC Name2-(dimethylamino)-1,3-thiazole-5-carbaldehyde
Molecular FormulaC6H8N2OS
InChI KeyZOPJEHPVWLTDJW-UHFFFAOYSA-N
SMILESCN(C)C1=NC=C(S1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Melting point72°C to 78°C
GC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only