Immuno Tubes and Stoppers
Immuno Tubes and Stoppers
Immuno Tubes and Stoppers
Immuno Tubes and Stoppers
Thermo Scientific™

Immuno Tubes and Stoppers

Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialDiameter (Metric) OuterProduct TypeFor Use With (Equipment)
444202Polystyrene12 mmImmuno Tubes and StopperImmuno Tube
341866Polyethylene11 mmStopper11 mm Nunc-Immuno Tubes
343036Polyethylene15 mmStopper15 mm Nunc-Immuno Tubes
348801Polyethylene12 mmStopper12 mm Nunc-Immuno Tubes
443990Polyethylene12 mmImmuno Tubes and StopperImmuno Tube
466982Polyethylene11 mmImmuno Tubes and Stopper
468608Polyethylene15 mmImmuno Tubes and StopperImmuno Tube
470319Polystyrene12 mmImmuno Star Tubes and StopperImmuno Tube
Catalog number 444202
Price (USD)
683.93
Online exclusive
844.60
Save 160.67
Case of 3000
Material:
Polystyrene
Diameter (Metric) Outer:
12 mm
Product Type:
Immuno Tubes and Stopper
For Use With (Equipment):
Immuno Tube
Price (USD)
683.93
Online exclusive
844.60
Save 160.67
Case of 3000

Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.

Features:
  • Offered in choice of three surfaces for optimal binding
  • Three tube sizes with stoppers available (ordered separately)
  • Fits standard equipment

Choice of Surface:

  • Thermo Scientific™ PolySorp™ is a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains.
  • Thermo Scientific™ MaxiSorp™ is a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character.
  • Thermo Scientific™ Mini-Sorp™ is a polyethylene-based material that provides a low-binding surface.

Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):

  • MiniSorp surface
  • Molded from special polyethylene formulation
  • For liquid phase immunoasssays, including RIA, for storing of reagents, or collecting chromatographic fractions

Solid Phase Immunoassay Tubes:

  • PolySorp and MaxiSorp surfaces available for choice of optimal binding
  • For solid phase immuno techniques such as IRMA, ELISA and ILMA
  • High–quality polystyrene for optimal quality
  • StarTube design gives higher sensitivity and faster assay times
  • Enhances assay sensitivity and reduces incubation times

Recommended for:

  • ELISA
  • Solid Phase RIA
  • Liquid Phase RIA
  • Sample storage
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Diameter (Metric) Outer12 mm
For Use With (Equipment)Immuno Tube
No. per Case3000
No. per Pack100
Length (Metric)75 mm
MaterialPolystyrene
Product TypeImmuno Tubes and Stopper
SurfaceMaxiSorp
Volume (Metric)5 mL
Unit SizeCase of 3000