Ethylene glycol, technical
Ethylene glycol, technical
Ethylene glycol, technical
Thermo Scientific Chemicals

Ethylene glycol, technical

CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Catalog NumberQuantity
4442300101 L
Catalog number 444230010
Price (USD)
36.10
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
36.10
Each
Chemical Identifiers
CAS107-21-1
IUPAC Nameethane-1,2-diol
Molecular FormulaC2H6O2
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILESOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
Appearance (Color)Clear colorless
Refractive index1.4300 to 1.4320 (20°C, 589 nm)
Water=<0.2 %
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only