Custom Plus TaqMan™ Copy Number Assay
Applied Biosystems™

Custom Plus TaqMan™ Copy Number Assay

Applied Biosystems TaqMan Copy Number Assays use gold-standard TaqMan MGB probe chemistry to evaluate the copy number of genomic DNARead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4442488L (2900 reactions), made to order
4442487S (360 reactions), made to order
4442520M (750 reactions), made to order
Catalog number 4442488
Price (USD)
1,764.00
Each
Quantity:
L (2900 reactions), made to order

Applied Biosystems TaqMan Copy Number Assays use gold-standard TaqMan MGB probe chemistry to evaluate the copy number of genomic DNA targets using Applied Biosystems real-time PCR instruments and software. TaqMan Copy Number Assays are run together with a TaqMan Copy Number Reference Assay in a duplex qPCR reaction; the copy number assay detects the target sequence, and the reference assay detects a sequence that is known to be present in two copies in the diploid genome. The results are then analyzed by the relative quantitation method using Applied Biosystems CopyCaller Software.

Custom Plus TaqMan Copy Number Assays are an optimal solution for studying variation in human and mouse genomic regions of interest for which a predesigned assay is not available. Using the free GeneAssist Copy Number Assay Tool, these assays are designed with the same bioinformatics pipeline used to develop the assays in the predesigned collection. The design process for Custom Plus assays includes up-front bioinformatic sequence analysis and in silico QC.

Benefits:
• Flexible—generate designs for human and mouse targets not covered by the predesigned assay collection using the free GeneAssist Copy Number Assay Tool
• Specific—gold-standard TaqMan chemistry with minor groove binder (MGB) probes increase assay specificity by enabling shorter probes
• Reproducible—robust assay designs developed with our bioinformatics pipeline enable accurate, reproducible, and reliable results
• Easy to interpret—CopyCaller Software provides the calculated copy number and predicted copy number, along with confidence value and z-score quality metrics
• Fast and simple—setup to primary analysis in 3–4 hours

Approximate ship time
4–6 days in North America and 6–10 days in Europe

All assay designs are the product of our industry-leading bioinformatics pipeline, optimized over the course of more than a decade by leveraging manufacturing and assay performance data. TaqMan Assays have been cited in over 40,000 publications and are backed by more than 350 patents.

TaqMan Copy Number Reference Assays are sold separately.

Recommended master mix (sold separately): TaqMan Genotyping Master Mix

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration60X
No. of Reactions2900 reactions
Product LineTaqMan
QuantityL (2900 reactions), made to order
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

Frequently asked questions (FAQs)

How can I reorder a Custom TaqMan Assay that I ordered previously?

To reorder the same Custom TaqMan Assay, you can follow these steps:
- Select an appropriate catalog number, considering the size of the assay that you want. You can find the different catalog numbers for Custom TaqMan Assays on their respective product pages on thermofisher.com.
- Once you have identified the necessary catalog number, go to www.thermofisher.com and click on "Quick Order", in the top right corner of the screen.
- In the following page, paste your selected catalog number in the "Catalog Number" field and paste the assay ID from your previous order in the "Assay ID" field.
- Click "Add to cart" and then in the following screen click "Proceed to check out".

Why do the values for the reference assay dye (VIC dye) vary for my TaqMan Copy Number Assay?

Please review the following possible causes and solutions:
-The reference sequence is not present or contains polymorphisms in the gDNA sample. Use an alternate copy number reference assay.
- There is variability in the amount of gDNA sample added to each reaction. Quantify the gDNA, then adjust the concentration of gDNA as needed.
Note: In general, the calculation of sample-level ΔCt accounts for variability in sample concentration.
- Pipetting was inaccurate. Check the pipette calibration of the pipettes, and pipette at least 5 µL of sample to prepare the reaction mix.

Why is the multicomponent signal for ROX dye not flat for my TaqMan Copy Number Assay?

Most likely incorrect dyes were selected for each target. Check the dyes selected for each target, then reanalyze the data.

Why was there a simultaneous increase in fluorescence from both the passive reference dye (ROX dye) and the reporter dyes for my TaqMan Copy Number Assay?

Most likely the sample evaporated. We recommend that you check the seal of the adhesive film for leaks before running the plate on the real-time PCR instrument.

Why was there a decrease in ROX dye fluorescence (passive reference dye) in my TaqMan Copy Number Assay?

Please review the following possible causes and solutions:
- There was insufficient Master Mix added to the reaction. Please follow accurate pipetting practices.
- Reagents are degraded. Ensure that the kits and reagents have been stored according to the instructions on the packaging and that they have not expired.