Replacement Lamp for ViiA™ 7 Real-Time PCR System

Catalog number: 4443978

Applied Biosystems™  Related applications: Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Replacement lamp, light source, for the ViiA™ 7 Instrument. Calibration is recommended after lamp replacement.

Specifications

For Use With (Equipment): ViiA™ 7 System
Product Line: Applied Biosystems™, ViiA™
Product Size: 1 lamp
Shipping Condition: Room Temperature
Type: Lamp

Contents & storage

Store at room temperature.

Documents

Manuals & protocols