Nitric acid, technical, 60% solution in water, Thermo Scientific Chemicals
Nitric acid, technical, 60% solution in water, Thermo Scientific Chemicals
Nitric acid, technical, 60% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitric acid, technical, 60% solution in water, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantityPackaging
4444000505 LPlastic drum
Catalog number 444400050
Price (USD)
-
Quantity:
5 L
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Titration with NaOH59 to 63 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only