4-Amino-2-chlorophenylboronic acid pinacol ester, 97%
4-Amino-2-chlorophenylboronic acid pinacol ester, 97%
4-Amino-2-chlorophenylboronic acid pinacol ester, 97%
4-Amino-2-chlorophenylboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

4-Amino-2-chlorophenylboronic acid pinacol ester, 97%

CAS: 877160-63-9 | C12H17BClNO2 | 253.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 444480010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS877160-63-9
IUPAC Name3-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular FormulaC12H17BClNO2
InChI KeyYKQXROMICPMQBZ-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=C(Cl)C=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point120°C to 125°C
Appearance (Color)Off-white to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only