Product Image
Applied Biosystems™

TaqMan™ Array Rodent MicroRNA A+B Cards Set v3.0

The TaqMan™ Array Rodent MicroRNA A+B Cards Set v3.0 is a two card set containing a total of 384 TaqMan™Read more
Have Questions?
Catalog NumberQuantity
44449094 x 2 cards
Catalog number 4444909
Price (USD)
3,880.00
Each
Quantity:
4 x 2 cards
Price (USD)
3,880.00
Each
The TaqMan™ Array Rodent MicroRNA A+B Cards Set v3.0 is a two card set containing a total of 384 TaqMan™ MicroRNA Assays per card. The set enables accurate quantitation of 641 and 373 unique microRNAs for mouse and rat respectively in as little as 5 hours. Included on each array is three TaqMan™ MicroRNA Assay endogenous controls for each species to aid in data normalization and one TaqMan™ MicroRNA Assay not related to rodent as a negative control. Detail on the included assays is available in the Target List file. Purchase of the Megaplex™ RT Primers, Rodent Pool Set v3.0 is required to run this array set. An additional preamplification step is enabled by Megaplex™ PreAmp Primers, Rodent Pool Set v3.0 for situations where sensitivity is of the utmost importance and/or sample is limiting.

TaqMan™ Array MicroRNA Cards provide all the advantages of TaqMan™ MicroRNA Assays in a convenient, pre-configured microfluidic card, minimizing the experimental variability and effort required to run 384 TaqMan™ MicroRNA Assays in parallel.

Megaplex™ RT Primers are highly multiplexed RT primers designed to convert up to 381 miRNAs to cDNA prior to real-time analysis, and are required to run the TaqMan™ Array MicroRNA Card. For greater sensitivity, or when working with limited sample, Megaplex™ PreAmp Primers provide an optional preamplification step prior to real-time analysis. TaqMan™ Array MicroRNA Cards and Megaplex™ Primer Pools enable a comprehensive expression profile consistent with miRBase v21 to be created in as little as 5 hours, providing the ideal miRNA profiling solution. Key product features:

Highly specific—quantitate only the biologically active mature miRNAs

• Highly streamlined workflow reduces the number of RT (reverse transcription) reactions per sample
• Simplify sample handling and increase sample throughput
• Coverage for human, mouse and rat species
• An ideal miRNA profiling solution

Simplify Sample Handling and Increase Sample Throughput
Following reverse transcription of miRNA targets using Megaplex™ RT Primers and optional preamplification using Megaplex™ PreAmp Primers, TaqMan™ Universal PCR Master Mix II is simply combined with each reaction and pipetted into each sample loading port in the TaqMan™ Array Card, simplifying sample handling and increasing sample throughput.

Provides Comprehensive Coverage of Sanger miRBase
Megaplex™ RT Pools and PreAmp Primers are available for human and rodent species, and consist of pre-selected and matched content across a series of primer pools and TaqMan™ Array Cards. The content is derived from miRBase v14 and v15 for human and rodent species respectively, providing close to full coverage of Sanger with the most up-to-date TaqMan™ MicroRNA Assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AB 7900 HT 384-Well System
FormDried Down
Includes384-Well Arrays, Information CD
Label or DyeFAM
Product LineTaqMan
Quantity4 x 2 cards
Reaction SpeedStandard
Research CategoryGene Regulation, Epigenetics
TypeArray Rodent MicroRNA A+B Cards Set
Format384-well Microfluidic Card
SpeciesMouse, Rat
Unit SizeEach
Contents & Storage
Contents: 384-well arrays and information CD. Storage: The TaqMan™ MicroRNA Arrays are shipped at ambient temperature. Upon arrival, store arrays at 2 to 8 ° and keep them protected from light.

Frequently asked questions (FAQs)

Why does the negative control well show amplification when doing microRNA analysis using Megaplex Primer Pools and TaqMan Array Cards?

Most likely the reagents or the cDNA template are contaminated. Please follow established PCR laboratory best practices.

Why do I have poor reproducibility across technical replicates when doing microRNA analysis using Megaplex Primer Pools and TaqMan Array Cards?

Most likely the reagents were not adequately mixed. Ensure that all samples and reagents are mixed well.

When doing microRNA analysis using Megaplex Primer Pools and TaqMan Array Cards, why are the Ct values for the endogenous controls highly variable across the sample set?

Highly variable values for endogenous controls is most likely due to biological variation. We recommend that you use an alternative endogenous control, such as a non-variable miRNA.

Why is the Ct value for the no-template control (NTC) <35 for some TaqMan microRNA Assays?

The Ct value for the NTC can be <35 for the following reasons:
- There are non-specific interactions between primers. For NTC information on a specific assay, see the Megaplex Assay Performance File.
- The cDNA template is contaminated. Please ollow established PCR good laboratory practices to prevent contamination.
- The preamplification product was not diluted properly before real-time PCR. Ensure that you dilute the preamplification product according the procedure in the user guide.

Why can I not detect any miRNA in my sample using the TaqMan MicroRNA Assay?

Most likely, the threshold is set too high to detect miRNA in samples with low levels of expression. We recommend adjusting the threshold (Ct) or NOAMP flag threshold (Crt) to an appropriate level.