TaqMan™ Array Human MicroRNA A+B Cards Set v3.0
Applied Biosystems™

TaqMan™ Array Human MicroRNA A+B Cards Set v3.0

The TaqMan® Array Human MicroRNA Card Set v3.0 is a two card set containing a total of 384 TaqMan® MicroRNARead more
Have Questions?
Catalog NumberQuantity
44449138 pack
Catalog number 4444913
Price (USD)
3,444.65
Online exclusive
3,870.00
Save 425.35
Each
Quantity:
8 pack
Price (USD)
3,444.65
Online exclusive
3,870.00
Save 425.35
Each
The TaqMan® Array Human MicroRNA Card Set v3.0 is a two card set containing a total of 384 TaqMan® MicroRNA Assays per card. The set enables accurate quantitation of 754 human microRNAs in as little as 5 hours. Included on each array is three TaqMan MicroRNA Assay endogenous controls to aid in data normalization and one TaqMan® MicroRNA Assay not related to human as a negative control. Details on the included assays is available in the Target List file. Purchase of the Megaplex™ RT Primers, Human Pool Set v3.0 is required to run this array set. An additional preamplification step is enabled by Megaplex™ PreAmp Primers, Human Pool Set v3.0 for situations where sensitivity is of the utmost importance and/or sample is limiting.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AB 7900 HT 384-Well System
FormDried Down
Label or DyeFAM
Product LineMegaplex
Quantity8 pack
Reaction SpeedStandard
Research CategoryGene Regulation, Epigenetics
TypeArray Human MicroRNA A+B Cards Set
Format384-well Microfluidic Card
SpeciesHuman
Unit SizeEach
Contents & Storage
4°C

Frequently asked questions (FAQs)

Why does the negative control well show amplification when doing microRNA analysis using Megaplex Primer Pools and TaqMan Array Cards?

Most likely the reagents or the cDNA template are contaminated. Please follow established PCR laboratory best practices.

Why do I have poor reproducibility across technical replicates when doing microRNA analysis using Megaplex Primer Pools and TaqMan Array Cards?

Most likely the reagents were not adequately mixed. Ensure that all samples and reagents are mixed well.

When doing microRNA analysis using Megaplex Primer Pools and TaqMan Array Cards, why are the Ct values for the endogenous controls highly variable across the sample set?

Highly variable values for endogenous controls is most likely due to biological variation. We recommend that you use an alternative endogenous control, such as a non-variable miRNA.

Why is the Ct value for the no-template control (NTC) <35 for some TaqMan microRNA Assays?

The Ct value for the NTC can be <35 for the following reasons:
- There are non-specific interactions between primers. For NTC information on a specific assay, see the Megaplex Assay Performance File.
- The cDNA template is contaminated. Please ollow established PCR good laboratory practices to prevent contamination.
- The preamplification product was not diluted properly before real-time PCR. Ensure that you dilute the preamplification product according the procedure in the user guide.

Why can I not detect any miRNA in my sample using the TaqMan MicroRNA Assay?

Most likely, the threshold is set too high to detect miRNA in samples with low levels of expression. We recommend adjusting the threshold (Ct) or NOAMP flag threshold (Crt) to an appropriate level.