2-Aminopyridine-4-boronic acid pinacol ester, 97%
2-Aminopyridine-4-boronic acid pinacol ester, 97%
2-Aminopyridine-4-boronic acid pinacol ester, 97%
Thermo Scientific Chemicals

2-Aminopyridine-4-boronic acid pinacol ester, 97%

CAS: 1195995-72-2 | C11H17BN2O2 | 220.08 g/mol
Have Questions?
Catalog NumberQuantity
4445800101 g
Catalog number 444580010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1195995-72-2
IUPAC Name4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular FormulaC11H17BN2O2
InChI KeyDCYKWKYBNWRLLZ-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC(N)=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
Melting point158°C to 164°C
Appearance (Color)White to tan
Infrared spectrumConforms
GC>=96 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only