Where can I download the software for a specific qPCR instrument?
To find the necessary software download for your instrument, please visit our Software, Patches, & Updates page and select your instrument so see relevant software downloads.