Product Image
Applied Biosystems™

SOLiD™ EZ Bead™ Emulsifier (110/220 V)

This is the Emulsifier component of the SOLiD™ EZ Bead™ System.
Have Questions?
Catalog NumberQuantity
44484191 Ea.
Catalog number 4448419
Price (USD)
6,500.00
1 instrument
Quantity:
1 Ea.
Price (USD)
6,500.00
1 instrument

This is the Emulsifier component of the SOLiD™ EZ Bead™ System. The Emulsifier combines an aqueous phase with an oil phase, creating a microreactor emulsion, bringing the necessary reagents and components for amplifying the library of interest onto a solid surface. This process takes up to 30 minutes to complete, depending on the desired bead yield.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Dimensions (L x W x H) Exterior40 cm x 28.1 cm x 33 cm
For Use With (Application)Next-Generation Sequencing
For Use With (Equipment)5500 Genetic Analyzer
Height (Metric)33 cm
High-throughput CompatibilityHigh-throughput Compatible, Multiplexing
Length (Metric)40 cm
LibrariesMate-Paired Library
Max. Voltage220 V, 110 V
Product TypeSOLiD EZ Bead Emulsifier
Quantity1 Ea.
Run Time30 min
Throughput20-30 GB⁄day downstream sequencing with 5500xl (microbeads), 30-45 GB⁄day (nanobeads)
Width (Metric)28.1 cm
Product LineEZ Bead, SOLiD
Unit Size1 instrument
Contents & Storage
The system includes:

• SOLiD EZ Bead Emulsifier
• 100 – 240 V power cable
• USB stick with documentation