Potassium ethyltrifluoroborate, 97%
Potassium ethyltrifluoroborate, 97%
Potassium ethyltrifluoroborate, 97%
Thermo Scientific Chemicals

Potassium ethyltrifluoroborate, 97%

CAS: 44248-07-9 | C2H5BF3K | 135.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 444880010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS44248-07-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Solid
Infrared spectrumConforms
Melting point>=260°C
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only