Pinacol éster de ácido 2,6-dimetoxipiridina-3-borónico, 97 %, Thermo Scientific Chemicals
Pinacol éster de ácido 2,6-dimetoxipiridina-3-borónico, 97 %, Thermo Scientific Chemicals
Pinacol éster de ácido 2,6-dimetoxipiridina-3-borónico, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pinacol éster de ácido 2,6-dimetoxipiridina-3-borónico, 97 %, Thermo Scientific Chemicals

CAS: 214360-59-5 | C13H20BNO4 | 265.12 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo 444920050
Precio (USD)
-
Cantidad:
5 g
Pedido a granel o personalizado
Identificadores químicos
CAS214360-59-5
IUPAC Name2,6-dimethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular FormulaC13H20BNO4
InChI KeyVZGDCNFVMWRPOW-UHFFFAOYSA-N
SMILESCOC1=CC=C(B2OC(C)(C)C(C)(C)O2)C(OC)=N1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Colorless to off-white
NMR (1H-NMR)Consistent with structure
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only