Veriti™ Dx 96-well Fast Thermal Cycler, 0.1 mL
Applied Biosystems™

Veriti™ Dx 96-well Fast Thermal Cycler, 0.1 mL

The Veriti™ Dx 96-Well Fast Thermal Cycler delivers the proven reliability you expect from Applied Biosystems for in vitro diagnosticRead more
Have Questions?
Catalog NumberQuantity
44522991 instrument
Catalog number 4452299
Price (USD)
20,080.00
Each
Quantity:
1 instrument
Price (USD)
20,080.00
Each
The Veriti™ Dx 96-Well Fast Thermal Cycler delivers the proven reliability you expect from Applied Biosystems for in vitro diagnostic PCR use. It features an easy-to-use color touch screen to simplify setup and use of the system, and VeriFlex™ Blocks, six independent temperature blocks designed to give precise control over your PCR optimization. Optional setups for fast PCR or standard PCR methods provide flexibility to shorten your PCR cycling time.

• Classified as US FDA Class I Medical Device for in vitro diagnostic use
• Conforms to IVDD (98⁄79⁄EC) requirements and labeled in Europe with a CE IVD mark
• Manufactured to ISO 13485 and GMP requirements
• Features an easy-to-use graphical interface (6.5 inch VGA touch screen)
• Innovative VeriFlex™ Blocks provide convenient PCR optimization
• Standard and fast-enabled cycling address your current and future PCR needs
• Reduced PCR reaction time when using faster ramp rates
• Comes with a 1-year warranty

The First End-Point Thermal Cycler Available for In Vitro Diagnostic Use
The Veriti™ Dx Thermal Cycler is a diagnostic instrument classified as a US FDA Class I Medical Device. It conforms to IVDD (98⁄79⁄EC) requirements and is CE IVD–labeled in Europe. The instrument is manufactured to ISO 13485 and GMP requirements.

Control at Your Fingertips
The powerful yet simple-to-operate user interface on the Veriti™ Dx 96-Well Fast Thermal Cycler is driven by the 6.5 inch (16.51 cm) VGA color touch screen. The large screen allows for easy viewing of your temperature profiles. Additionally, the large navigation buttons put programming of the Veriti™ Dx Thermal Cycler at your fingertips. Setup and navigation of the Veriti™ Dx Thermal Cycler do not require use of a stylus or mouse.

VeriFlex™ Blocks: Enhanced PCR Functionality
The Veriti™ Dx 96-Well Fast Thermal Cycler features innovative VeriFlex™ Blocks for enhanced PCR functionality. Six separate peltier blocks provide maximal versatility and flexibility and offer two key benefits: 1) PCR optimization—the ability to set each block at a specific temperature is ideal for precise control over PCR optimization; 2) run more experiments: the blocks can also be used for up to six different annealing temperatures in the same run. With this enhanced functionality, the Veriti™ Dx 96-Well Fast Thermal Cycler with VeriFlex™ Blocks is the ideal thermal cycler for any lab.

Fast Setup, Fast Results
The Veriti™ Dx 96-Well Fast Thermal Cycler has two different options for method navigation. For quick setup, you may select one of the preprogrammed run methods. If you prefer to program your own methods, simply touch the step you would like to set up, and then use the keypad to enter thermal cycling steps. After completing a setup, you have the choice of saving the method to the instrument or to a USB memory stick.

Control Your Time: Go Fast When You Want
With the Veriti™ Dx 96-Well Fast Thermal Cycler you get two thermal cyclers in one: a standard thermal cycler with its better-than-gradient feature, and a fast-enabled thermal cycler. So whether you have a current need to run fast PCR and shorten your PCR cycling times, or a possible future application need for fast PCR, the Veriti™ Dx Thermal Cycler provides the flexibility to go fast when you want.
Specifications
Dimensions19.1 x 9.3 x 9.6 in. (48.5 x 23.7 x 24.5 cm)
Display Type6.5 in. VGA 32 k Color Touchscreen
For Use With (Equipment)Veriti Dx Thermal Cycler
Max. Ramp Rate5°C/sec. (Block), 4.25°C/sec. (Sample)
Quantity1 instrument
Reaction SpeedFast, Standard
Thermal Accuracy±0.25°C (35°C to 99.9°C)
Thermal Range0°C to 100°C
Thermal Uniformity<0.5°C (20 sec. after reaching 95°C)
User InterfaceUSB, Wi-Fi, Ethernet
Voltage100/240 V
WarrantyLimited Warranty
Weight25 lb. (11.4 kg)
Block Configurations96-well Fast, 6-Zone VeriFlex Block
Block FormatNon-interchangeable
Capacity96-well, 0.1 mL
No. of Programs>500
Product TypeThermal Cycler
Unit SizeEach

Frequently asked questions (FAQs)

What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf

Find additional tips, troubleshooting help, and resources within ourPCR Thermal Cyclers Support Center.