The 5500 Genetic Analyzer, Catalog # 4452865 has been discontinued. The Ion Torrent S5 Sequencers are available as alternatives to this product. View discontinuation letter ›
5500 Genetic Analyzer
Applied Biosystems™

5500 Genetic Analyzer

Built on three decades of sequencing expertise and powered by the proven accuracy of ligation-based sequencing, the new 5500 GeneticRead more
Have Questions?
Catalog NumberQuantity
44528651 instrument
Catalog number 4452865
Price (USD)
-
Quantity:
1 instrument
Built on three decades of sequencing expertise and powered by the proven accuracy of ligation-based sequencing, the new 5500 Genetic Analyzer provides a robust, affordable next-generation sequencing platform for every lab. Designed to give you the choice to optimize for turnaround time or throughput, the 5500 Genetic Analyzer is a highly accurate, cost-effective solution for analyzing one or many samples.

Key Benefits
• Sequencing right here, right now
• Cost-effective sequencing runs with individually configurable lanes
• Superior low frequency variant detection for whole exome sequencing or targeted resequencing
• Reproducibility, reliability, and quality for your RNA applications
• Confidence and uniformity in multiplexing with up to 96 barcodes
• Optimal analysis solutions for streamlined productivity and freedom to operate

Explore the Depths of Translational Research
With Exact Call Chemistry, the industry-leading accuracy of the 5500 Genetic Analyzer helps you get definitive answers about biological variation for applications like whole exome, RNA-seq, and structural variation.

Customize the System for Your Projects
The 5500 Genetic Analyzer allows you to tailor your sequencing runs precisely for your research. The six independent FlowChip lanes and Pay-Per-Lane Sequencing (PPL-Seq™) allow you to configure the instrument to different project scales and application types. Additionally, the system’s intelligent barcode kits can accommodate from one to hundreds of samples in a single run.

Don’t Wait
With the 5500 Genetic Analyzer, the unique capability to run each FlowChip lane independently allows you to turn around results in the shortest time possible. Now you can explore the genome, transcriptome, and epigenome with a hypothesis-neutral platform that combines powerful sequence data generation with premium accuracy. Whether you're running whole exome, RNA-seq, or a structural variation experiment, the 5500 Genetic Analyzer is designed to deliver high-quality results in a time-sensitive and cost-effective manner.

Perform Sequencing with Ease
Designed in collaboration with core facilities and principal investigators, the integrated 5500 Genetic Analyzer provides an intuitive and user-friendly workflow from sample preparation to data analysis.

Workflow Benefits Include:
• Embedded sequencing controls to enable real-time monitoring of data quality throughout the entire workflow
• System alerts to help maximize data quality and system performance, and to track reagent usage during the course of the sequencing run
• Easy-to-use software, 60% smaller data footprints, workstation computing, and standard base sequence file formats to facilitate setup, daily operations, and seamless integration with data analysis tools

Join the Era of Genomic Medicine
The throughput, broad range of applications, system flexibility, and intuitive design of the 5500 Genetic Analyzer address the needs of core and services laboratories, and basic and clinical research labs, to take sequencing into every scientific discipline.

A Community of Support
With more than 2,000 field personnel, we have the most extensive network of dedicated Field Application Specialists and Service Engineers in the industry. You can feel confident that we will be there when and where you need us most.

The 5500 Genetic Analyzer is For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accuracy0.9999
Data Outputs.bam files, .xsq files
Dimensions (L x W x H) Exterior120 mm x 750 mm x 800 mm
DisplayGUI
Electrical Requirements240 V
External ComputerTower Computer
For Use With (Application)Next-Generation Sequencing
For Use With (Equipment)5500 Genetic Analyzer
Height (English) Exterior31.4 in.
Height (Metric) Exterior800 mm
High-throughput CompatibilityMultiplexing
Length (English) Exterior4.72 in.
Length (Metric) Exterior120 mm
LibrariesFragment Library, Mate-Paired Library, Paired-End Library
Max. Voltage240V
Number of Lanes6 lanes
Quantity1 instrument
Read Length75 bp (fragment), 75 bp x 35 bp (paired-end), Up to 60 bp x 60 bp (mate-paired)
Run Time24 hrs
Sequencing Scalegenome, exome, transcriptome
ThroughputUp to 10-15 Gb per day
Width (English) Exterior29.5 in.
Width (Metric) Exterior750 mm
Product LineSOLiD
Unit Size1 instrument
Contents & Storage
Altitude:
Located between sea level and 2000m (6500 ft.) above sea level

Humidity:
Sequencer, computer, and the UPS unit
20%–80% (noncondensing)

Temperature:
Sequencer
15–24°C
Note: The room temperature must not fluctuate more than 2°C over a 2-hour period.