ViiA™ 7 Dx Instrument, array card, desktop - FAQs
ViiA™ 7 Dx Instrument, array card, desktop - FAQs
View additional product information for ViiA™ 7 Dx Instrument, array card, desktop - FAQs (4453542)
1 product FAQs found
Is the Maxima SYBR Green/ROX qPCR Master Mix (2X) (Cat. No. K0223, K0221, K0222) compatible with the ViiA 7 Real-Time PCR System (Cat. No. 4453542, 4453539)
No, the ViiA 7 Real-Time PCR System (Cat. No. 4453542, 4453539) is not compatible with the Maxima SYBR Green/ROX qPCR Master Mix (2X) (Cat. No. K0223).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.