Product Image
Applied Biosystems™

ViiA™ 7 System Block Upgrade Kit, TaqMan™ Array

Allows the ViiA™ 7 Real-Time PCR System to operate using a TaqMan™ Array Block.
Have Questions?
Catalog NumberQuantity
44535461 upgrade
Catalog number 4453546
Price (USD)
-
Quantity:
1 upgrade
This kit allows the ViiA™ 7 Real-Time PCR System to operate using a TaqMan™ Array Block.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)ViiA™ 7 System
Format384-well Microfluidic Card
Product LineTaqMan
Quantity1 upgrade
Reaction SpeedStandard
Capacity1 Array Card
Product TypeSystem Block Upgrade Kit
Unit Size1 upgrade
Contents & Storage
Includes TaqMan Array Block, heated cover, plate adaptor and calibration kit.

Frequently asked questions (FAQs)

I cannot open the QuantStudio 3/5 Design & Analysis 2 (DA2) software on my Mac. What are the software requirements for Mac?

To open the software download on your Mac, you will need to adjust your security settings using the following instructions from Apple support:

Safely open apps on your mac

Please note, the software should run on Mac OS 10.10 or later.

Find additional tips, troubleshooting help, and resources within our QuantStudio 3 and QuantStudio 5 Real Time PCR Systems Support Center.

Can I import a standard curve for my real-time PCR data into the Design & Analysis Software (v2.x)?

It is possible to import a standard curve into the Design & Analysis Software (v2.x) if you are analyzing results from one of the following Applied Biosystems real-time PCR instruments:
• 7900HT Fast Real-Time PCR System
• 7500/7500 Fast Real-Time PCR System
• StepOne/StepOne Plus Real-Time PCR System
• ViiA 7 Real Time PCR System
• QuantStudio 1 Real-Time PCR System
• QuantStudio 3 & 5 Real-Time PCR System
• QuantStudio 6 & 7 Flex Real-Time PCR System
• QuantStudio 6 & 7 Pro Real-Time PCR Systems
• QuantStudio 12K Flex Real-Time PCR system

Follow the instructions below to import a standard curve for analysis in the Design & Analysis Software (v2.x):
1. Open the data file in the Design & Analysis Software (v2.x).
2. Click on Actions.
3. Click on "Standard Curve Analysis Setting".
4. Select "External Standard Curves" and click on "Import".
5. Browse to the proper .csv file and click "Open".
6. Click "Apply".

Can I import a standard curve for my real-time PCR data into the QuantStudio Real-Time PCR Software?

If you are analyzing results from the ViiA 7 Real-Time PCR System, QuantStudio 6 Flex Real-Time PCR System, or QuantStudio 7 Flex Real-Time PCR System, it is possible to import a standard curve into the QuantStudio Real-Time PCR Software following the instructions below:
1. Open the data file in the QuantStudio Real-Time PCR Software.
2. Go to Analysis Settings.
3. Click on the "Standard Curve Settings" tab.
4. Select the option to "Use a standard curve imported from another experiment" and click on "Import".
5. Browse to and highlight the proper data file and click "Select".
6. Click Apply Analysis Settings.

Is the ViiA7 Real-Time PCR System compatible with the 7900HT Fast System Twister robot?

No, the ViiA7 Real-Time PCR System is not compatible with the 7900HT Fast System Twister robot. For automation, the ViiA7 Real-Time PCR System can be connected to one of the following robots:
• Orbitor RS2 Microplate Mover for QuantStudio Systems, Sequential Access (cat. No. A32268)
• Orbitor RS2 Microplate Mover for QuantStudio Systems, Random Access (cat. No. A32269)
• Twister Automation Robot for QuantStudio Systems (Cat. No. 4471066)

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

How can I get a summary of the calibrations performed on the QuantStudio 6 and 7 Flex Real-Time PCR Systems?

You can get a summary of the calibrations from the QuantStudio Software by going to Instrument > Instrument Console > Manage Instrument > Information > Print Calibration Log.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.