2,4-Dimethoxypyrimidine-5-boronic acid, 95%
2,4-Dimethoxypyrimidine-5-boronic acid, 95%
2,4-Dimethoxypyrimidine-5-boronic acid, 95%
2,4-Dimethoxypyrimidine-5-boronic acid, 95%
Thermo Scientific Chemicals

2,4-Dimethoxypyrimidine-5-boronic acid, 95%

CAS: 89641-18-9 | C6H9BN2O4 | 183.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 445420050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS89641-18-9
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Additional infoMay contain varying amounts of anhydrides
NMR (1H-NMR)Consistent with structure
Appearance (Color)White to off-white
Appearance (Form)Solid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only