4-[(Ethoxycarbonyl)methyl]phenylboronic acid, 97%
4-[(Ethoxycarbonyl)methyl]phenylboronic acid, 97%
4-[(Ethoxycarbonyl)methyl]phenylboronic acid, 97%
4-[(Ethoxycarbonyl)methyl]phenylboronic acid, 97%
Thermo Scientific Chemicals

4-[(Ethoxycarbonyl)methyl]phenylboronic acid, 97%

CAS: 92243-74-8 | C10H13BO4 | 208.02 g/mol
Have Questions?
Catalog NumberQuantity
4454300101 g
Catalog number 445430010
Price (USD)
495.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
495.00
Each
Chemical Identifiers
CAS92243-74-8
IUPAC Name[4-(2-ethoxy-2-oxoethyl)phenyl]boronic acid
Molecular FormulaC10H13BO4
InChI KeyRDIRUZGSOKLANG-UHFFFAOYSA-N
SMILESCCOC(=O)CC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Additional infoMay contain varying amounts of anhydrides
Appearance (Form)Solid
Appearance (Color)Off-white to tan
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only