Phenylboronic acid neopentylglycol ester, 97%
Phenylboronic acid neopentylglycol ester, 97%
Phenylboronic acid neopentylglycol ester, 97%
Phenylboronic acid neopentylglycol ester, 97%
Thermo Scientific Chemicals

Phenylboronic acid neopentylglycol ester, 97%

CAS: 5123-13-7 | C11H15BO2 | 190.05 g/mol
Have Questions?
Catalog NumberQuantity
4455000505 g
Catalog number 445500050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS5123-13-7
IUPAC Name5,5-dimethyl-2-phenyl-1,3,2-dioxaborinane
Molecular FormulaC11H15BO2
InChI KeyFQMSNYZDFMWLGC-UHFFFAOYSA-N
SMILESCC1(C)COB(OC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Solid
Appearance (Color)White
Melting point62°C to 67°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only