Product Image
Applied Biosystems™

Attune™ Flow Cytometer Focusing Fluid Filter

This 0.2 micron filter prevents contaminants from getting into the focusing fluid reservior of the Attune Flow Cytometer.
Have Questions?
Catalog NumberQuantity
4456564Each
Catalog number 4456564
Price (USD)
111.65
Online exclusive
126.00
Save 14.35
Each
Quantity:
Each
Price (USD)
111.65
Online exclusive
126.00
Save 14.35
Each

This 0.2 micron filter prevents contaminants from getting into the focusing fluid reservior of the Attune Flow Cytometer. Recommended for replacement once a month.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Product LineAttune
QuantityEach
Shipping ConditionRoom Temperature
For Use With (Equipment)Attune Acoustic Focusing Cytometer
Product TypeFocusing Fluid Filter
Unit SizeEach
Contents & Storage
Store at room temperature