Tetrachloroethylene, 99+%, for HPLC
Tetrachloroethylene, 99+%, for HPLC
Tetrachloroethylene, 99+%, for HPLC
Tetrachloroethylene, 99+%, for HPLC
Thermo Scientific Chemicals

Tetrachloroethylene, 99+%, for HPLC

CAS: 127-18-4 | C2Cl4 | 165.82 g/mol
数量:
100 mL
1 L
2.5 L
パッケージ:
Glass bottle
製品番号(カタログ番号) 445691000
価格(JPY)
-
見積もりを依頼する
数量:
100 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS127-18-4
IUPAC Nametetrachloroethene
Molecular FormulaC2Cl4
InChI KeyCYTYCFOTNPOANT-UHFFFAOYSA-N
SMILESClC(Cl)=C(Cl)Cl
さらに表示
Appearance (Form)Clear liquid
GC>=99.0 %
Residue after evaporation=<0.0005 %
UVat 295 nm A: =<0.3
UVat 400 nm A: =<0.03
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only