Ambion™ In Vivo Pre-Designed siRNA HPLC IVR, 1 μmol - FAQs
Ambion™ In Vivo Pre-Designed siRNA HPLC IVR, 1 μmol - FAQs
View additional product information for Ambion™ In Vivo Pre-Designed siRNA - FAQs (4457303, 4457310, 4457301, 4457302, 4457307, 4457308, 4457309)
2 product FAQs found
What are the benefits of using our Invitrogen In vivo siRNA?
- 100x more stable than unmodified siRNA - longer knockdown in vivo
- Compatible with any 21-mer sequence - no loss of potency
- Non-toxic - actually reduces siRNA sequence specific immune responses
- Enhanced specificity - up to 90% reduction in off target effects
- Both Pre-designed and Validated Invitrogen In Vivo siRNAs offered
Find additional tips, troubleshooting help, and resources within our RNAi Support Center.
What siRNA do you offer that I can use for my in vivo studies, and what are the differences between them?
Please see the different siRNA we offer for in vivo studies:
Ambion In Vivo Pre-designed siRNAs are designed using the Silencer Select algorithm and incorporate additional chemical modifications for superior serum stability (half life >5 hours at 37 degrees C in 90% mouse serum) with in vivo applications. Ambion In Vivo siRNAs are non-toxic and non-immunogenic in vitro (peripheral blood nononuclear cells; PBMC) and in vivo (mouse). In cell-based assays, Ambion In Vivo siRNAs exhibit potency and specificity equivalent to Silencer Select siRNAs.
Ambion In Vivo Custom Designed siRNAs are designed to your specified target, using the Silencer Select algorithm and Ambion In Vivo chemical modifications.
Ambion In Vivo Custom Synthesized siRNAs are synthesized with your sequence, with Ambion In Vivo chemical modifications for serum stability.
Our Ambion In Vivo siRNA can be ordered with the following purities:
Desalted: Desalted and analyzed by MALDI-TOF mass spectrometry.
HPLC: HPLC purified and analyzed by MALDI-TOF and analytical HPLC
In Vivo Ready : HPLC purified, analyzed by MALDI-TOF and analytical HPLC, dialyzed to remove salts, sterile filtered, and endotoxin tested
Use our GeneAssist tool to order your custom siRNA (http://www.thermofisher.com/order/custom-genomic-products/tools/sirna/).
Find additional tips, troubleshooting help, and resources within our RNAi Support Center.