2-Ethoxyethanol, for analysis
2-Ethoxyethanol, for analysis
2-Ethoxyethanol, for analysis
Thermo Scientific Chemicals

2-Ethoxyethanol, for analysis

CAS: 110-80-5 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 445760025
Price (EUR)
170,00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (EUR)
170,00
Each
Chemical Identifiers
CAS110-80-5
IUPAC Name2-ethoxyethan-1-ol
Molecular FormulaC4H10O2
InChI KeyZNQVEEAIQZEUHB-UHFFFAOYSA-N
SMILESCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Magnesium (Mg)=<0.2 ppm
Color scale=<10 APHA
GC>=98.5 %
Acidity/alkalinity=<0.001 meq/g
Iron (Fe)=<0.5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only