1,1,1,3,3,3-Hexafluoro-2-propanol, 99%, for analysis
1,1,1,3,3,3-Hexafluoro-2-propanol, 99%, for analysis
1,1,1,3,3,3-Hexafluoro-2-propanol, 99%, for analysis
Thermo Scientific Chemicals

1,1,1,3,3,3-Hexafluoro-2-propanol, 99%, for analysis

CAS: 920-66-1 | C3H2F6O | 168.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Packaging:
View all
Glass Bottle
Catalog number 445822500
Price (USD)
975.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,148.00
Save 172.35 (15%)
Each
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
975.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,148.00
Save 172.35 (15%)
Each
Chemical Identifiers
CAS920-66-1
IUPAC Name1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular FormulaC3H2F6O
InChI KeyBYEAHWXPCBROCE-UHFFFAOYSA-N
SMILESOC(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Calcium (Ca)=<0.5 ppm
Water=<0.05 %
Copper (Cu)=<0.02 ppm
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only