Single Cell Lysis Kit
Single Cell Lysis Kit
Single Cell Lysis Kit
Invitrogen™

Single Cell Lysis Kit

Single cell analysis has the potential to transform life science research. However, detecting and quantitating the minute amounts of RNARead more
Have Questions?
Catalog NumberQuantity
4458235100 preps
Catalog number 4458235
Price (USD)
1,054.00
Each
Quantity:
100 preps
Price (USD)
1,054.00
Each
Single cell analysis has the potential to transform life science research. However, detecting and quantitating the minute amounts of RNA contained in single cells remains a significant challenge. The Ambion™ Single Cell Lysis Kit enables reliable RNA preparation for single cells.

Key product features:

• Optimized sample prep—Enables RNA preparation for single cells
• Efficient design—No sample loss ensures maximal sensitivity
• Versatile application—Compatible with common enzymatic reactions

Optimized RNA Sample Prep for 1-10 Cells
The Ambion™ Single Cell Lysis Kit enables RNA preparation from 1-10 cells. The Single Cell Lysis kit generates a concentrated cell lysate through the optimized combination of lysis buffer, stop solution, and DNase. The resulting lysate is compatible with common enzymatic reactions such as reverse transcription. Sample preparation is fast and can be completed in only 7 minutes with 2 pipetting steps (Figure 1).

No Sample Loss Ensures Maximal Sensitivity
The Ambion™ Single Cell Lysis Kit bypasses the need for traditional RNA purification. There are no sample transfers, columns or washes which can lead to loss and⁄or dilution of the precious sample. The entire lysate can be used in downstream reactions ensuring maximal sensitivity and accuracy in your experiments.

Compatible with Common Enzymatic Reactions
The reagents in the Ambion™ Single Cell Lysis Kit are identical to the lysis reagents in the complete Single Cell-to-CT™ Kit for Real-time RT-PCR analysis from 1-10 cells. They have been validated with multiple reverse transcription kits including SuperScript™ VILO™ cDNA Synthesis Kit, High Capacity RNA-to-cDNA kit and TaqMan™ MicroRNA Reverse Transcription Kit, for analysis of mRNA as well as miRNA targets. As a stand alone module, the Single Cell Lysis Kit allows you to create new workflows to expand the possibilities of single cell analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeLysate
High-throughput CompatibilityNot High-throughput Compatible (Manual)
No. of Reactions100 preps
Product LineAmbion, Cells-to-CT
Purification Time7 min.
Quantity100 preps
Sample TypeBacteria, Blood, Cells, Tissue, Viral Samples
Shipping ConditionDry Ice
Starting Material1-10 cells
TargetLysate, ready for real-time PCR
Concentration2X
For Use With (Application)Real Time PCR (qPCR), RT-PCR
Unit SizeEach
Contents & Storage
Kit contains:
  • 100 μL Single Cell Dnase I: -20°C;
  • 1 mL Single Cell Lysis Solution: 4°C;
  • 100 μL Single Cell Stop Solution: -20°C

Frequently asked questions (FAQs)

What is the minimum amount of cells required to perform RNA isolation using the Single Cell Lysis Kit?

The Single Cell Lysis Kit enables RNA preparation from 1-10 cells. It generates a concentrated cell lysate through the optimized combination of lysis buffer, stop solution, and DNase. The resulting lysate is compatible with common enzymatic reactions such as reverse transcription. Sample preparation is fast and can typically be completed in around 7 minutes with 2 pipetting steps.

Can laser micro-dissected FFPE single cells be used with a Single Cell-to-CT Kit?

While we have not tried this in-house, there is reason to believe this should work. However, optimization of the protocol around the RT step may be needed, as FFPE samples have a lot of RNA crosslinking, which could affect the efficiency of the RT enzyme. Heating the sample for 10 min at 80 degrees C before adding the RT in order to get good cDNA conversion should improve reverse transcription.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.