ABY™ Dye Spectral Calibration Plate, 96-well
Applied Biosystems™

ABY™ Dye Spectral Calibration Plate, 96-well

This plate is used for ABY spectral calibration of your Applied Biosystems 7500 or ViiA 7 real-time PCR system withMás información
Have Questions?
Número de catálogoCantidad
44615911 Plate
Número de catálogo 4461591
Precio (MXN)
-
Cantidad:
1 Plate
This plate is used for ABY spectral calibration of your Applied Biosystems 7500 or ViiA 7 real-time PCR system with a 96-well block. Please see your instrument maintenance guide for full instructions on use. The dye formulations will help improve multiplexing results by more accurately representing fluorescent spectra that are part of your real-time PCR experiments.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Tinte calibradoABY™
Tipo de calibraciónCalibración espectral
Para utilizar con (aplicación)PCR en tiempo real (PCRq)
Para utilizar con (equipo)Sistema 7500
Cantidad1 Plate
Condiciones de envíoHielo seco
Tipo de productoPlaca de calibración espectral de colorantes
Unit SizeEach
Contenido y almacenamiento
Almacenar en el congelador (de -5 °C a -30 °C).

Preguntas frecuentes

What are the excitation and emission profiles for ABY, JUN, Cyanine 5, and Cyanine 5.5 dyes?

ABY dye has an excitation wavelength of 568 nm and emission wavelength of 583 nm.
JUN dye has an excitation wavelength of 606 nm and emission wavelength of 618 nm.
Cynanine 5 dye has an excitation wavelength of 649 nm and emission wavelength of 667 nm.
Cyanine 5.5 dye has an excitation wavelength of 678 nm and emission wavelength of 695 nm.

Please refer to your instrument user guide for additional information on compatibility with different dye channels.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.