MUSTANG PURPLE™ Dye Spectral Calibration Plate, 96-well - FAQs
MUSTANG PURPLE™ Dye Spectral Calibration Plate, 96-well - FAQs
View additional product information for MUSTANG PURPLE™ Dye Spectral Calibration Plate, 96-well - FAQs (4461599)
1 product FAQs found
Do you offer MUSTANG PURPLE Dye Spectral Calibration Plate, 96-well (Cat. No. 4461599) as a stand-alone passive reference dye for purchase?
No, we do not offer Mustang Purple as a stand-alone product.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.