Acetic acid, sodium salt trihydrate, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Acetic acid, sodium salt trihydrate, for HPLC, Thermo Scientific Chemicals

Green features
Have Questions?
Catalog NumberQuantity
446202500250 g
Catalog number 446202500
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS6131-90-4
IUPAC Namesodium acetate trihydrate
Molecular FormulaC2H9NaO5
InChI KeyAYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Insoluble matter=<0.0005 %
pH7.5 to 9.2
Appearance (Color)White
UVat 254 nm A: =<0.02
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only