TaqMan™ OpenArray™ Real-Time PCR Master Mix, 1.5 mL - FAQs
TaqMan™ OpenArray™ Real-Time PCR Master Mix, 1.5 mL - FAQs
View additional product information for TaqMan™ OpenArray™ Real-Time PCR Master Mix - FAQs (4462164, 4462159)
1 product FAQs found
Which panel can the TaqMan OpenArray Real-Time PCR Master Mix be used with?
The Master Mix has been optimized for use with the TaqMan OpenArray gene expression panels, and yields reproducible, specific and sensitive results.