Sodium phosphate, monobasic dihydrate, for HPLC
Sodium phosphate, monobasic dihydrate, for HPLC
Sodium phosphate, monobasic dihydrate, for HPLC
Thermo Scientific Chemicals

Sodium phosphate, monobasic dihydrate, for HPLC

CAS: 13472-35-0 | H6NaO6P | 156.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
446222500250 gglass bottle
Catalog number 446222500
Price (USD)
-
Quantity:
250 g
Packaging:
glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13472-35-0
IUPAC Namesodium dihydrate dihydrogen phosphate
Molecular FormulaH6NaO6P
InChI KeyVBJGJHBYWREJQD-UHFFFAOYSA-M
SMILESO.O.[Na+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 254 nm A: =<0.05
pH4.0 to 4.5
Appearance (Color)White
Titration Acid98.5 to 101.5 %
Water=<24 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only