Matrix™ Capping Systems, SepraSeal, gray - FAQs
Matrix™ Capping Systems, SepraSeal, gray - FAQs
View additional product information for Matrix™ SepraSeal and DuraSeal Capping Systems - FAQs (4465RED, 4463PUR, 4464GRY, 4463RED, 4465GRE, 4463GRE, 4490, 4466YEL, 4464YEL, 4466, 4466BLU, 4464BLU, 4465PUR, 4466GRY, 4463GRY, 4464RED, 4466RED, 4463, 4464, 4465, 4466GRE, 4464GRE, 4463YEL, 4465YEL, 4465GRY, 4464PUR, 4465BLU, 4463BLU, 4466PUR)
2 product FAQs found
What are the main advantages of the Thermo Scientific 2D barcoded storage system?
Here are some advantages of the 2D barcoded storage system:
- Less time needed for labeling, sample recording, and retrieval
- Maximize freezer utilization while keeping storage space organised
- Ensures a safe, permanent ID of a sample
- Enables multivial capping and multichannel filling
- Individualized samples, eliminating multiple freeze/thaw cycles
Which Thermo Scientific 2D barcoded storage tubes should I use for my application?
Please view the following information regarding the usage of Thermo Scientific 2D barcoded storage tubes:
Tube Type; Typical Storage Temperature; Application(s); Special Feature(s)
Sepraseal; -20 degrees C; Compound storage; Can easily be removed for sample access
Presplit Sepraseal; -20 degrees C; Compound storage; Up to 200 piercings
Duraseal; -20 degrees; Compound storage; Safe seal for multiple piercings
Matrix ScrewTop; -80 degrees C; Blood components, DNA, Bacteria; With the correct torque, there is very little evaporation at -80 C, disposable pipette tips can be used
Nunc Screwcap; -176 degrees C; Mammalian cells; Minimal height, tubes can be removed with cryo gloves