Immuno Breakable Modules Clear, C8 LockWell, MediSorp
Immuno Breakable Modules Clear, C8 LockWell, MediSorp
Immuno Breakable Modules Clear, C8 LockWell, MediSorp
Immuno Breakable Modules Clear, C8 LockWell, MediSorp
Immuno Breakable Modules Clear, C8 LockWell, MediSorp
Thermo Scientific™

Immuno Breakable Modules Clear, C8 LockWell, MediSorp

Choose from four surfaces and a variety of wells and configurations. Immuno Breakable Modules offer an optimal choice of products for the specific assay.
Have Questions?
Catalog NumberSurface Treatment
446470MediSorp
Catalog number 446470
Price (USD)
506.69
Online exclusive
664.83
Save 158.14
Case of 60
Surface Treatment:
MediSorp
Price (USD)
506.69
Online exclusive
664.83
Save 158.14
Case of 60

Choose from four surfaces and a variety of wells and configurations. Thermo Scientific™ Immuno Breakable Modules offer an optimal choice of products for the specific assay. They offer lot-to-lot reliability and well-to-well reproducibility.

Choose between LockWell, BreakApart and Breakable 1x8 modules in standard 96-well frames.
  • For solid phase immuno assays, e.g. ELISA
  • Choice of four surfaces for optimal binding
  • Free-standing modules are easy to handle
  • Alphanumeric system for easy orientation
  • Fit standard equipment
  • High optical quality for reliable results
  • Polystyrene

Choice of Surface

  • PolySorp™, Immulon™ 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
Binding PropertySlightly Hydrophilic
BreakapartBreakable Strips
CertifiedYes
DescriptionHigh optical quality for reliable results
Detection MethodColorimetric
For Use With (Application)For binding a diverse range of biomolecules
LidWithout Lid
No. per Case60
No. per Pack10
Packaging60 Cases
Plate BlockingNone
Product TypeFrame
Target MoleculeProteins
Volume (Metric) Working250 μL
Array8 x 12
ColorClear
MaterialPolystyrene
No. of Wells96
Surface TreatmentMediSorp
Well ShapeRound, Curved Edges, Flat Bottom (C-well)
Unit SizeCase of 60

Frequently asked questions (FAQs)

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with LockWell modules?

Due to the locking feature design of the LockWell modules, the 8-Well Strip Caps for Immuno Standard Modules are not compatible with, and will not seal, the LockWell Modules.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.