Immuno Breakable Modules Clear
Actual product may vary
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Thermo Scientific™

Immuno Breakable Modules Clear

Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface TreatmentWell ShapeNo. per PackProduct Type
446477PolySorpRound10Frame
446442PolySorpRound, C Bottom5Strip Plate
446469MaxiSorpRound, C Bottom10Strip Plate
446490MultiSorpRound, C Bottom10Frame
446639MaxiSorpRound10Microplate Strip
473539PolySorpRound, C Bottom5Microplate Strip
473768MaxiSorpRound, C Bottom5Strip Plate
95029390Universal BindingRound, Flat Bottom (F-well)50Microplate Strip
Catalog number 446477
Price (USD)
-
Surface Treatment:
PolySorp
Well Shape:
Round
No. per Pack:
10
Product Type:
Frame

Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.

  • Free-standing modules are easy to handle
Specifications
Binding PropertyHydrophobic
BreakapartBreakable Strips
CertifiedNo
DescriptionHigh optical quality for reliable results
Detection MethodColorimetric
For Use With (Application)For adsorption of hydrophobic molecules
FormatStrip Plate
No. per Case60
No. per Pack10
Packaging10 Pk.
Plate BlockingNone
Product TypeFrame
Target MoleculeLipids
Volume (Metric) Working250 μL
ColorClear
MaterialPolystyrene
No. of Wells96
Surface TreatmentPolySorp
Well ShapeRound
Unit SizeEach

Frequently asked questions (FAQs)

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with LockWell modules?

Due to the locking feature design of the LockWell modules, the 8-Well Strip Caps for Immuno Standard Modules are not compatible with, and will not seal, the LockWell Modules.