Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Thermo Scientific™

Immuno Breakable Modules Clear

Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface TreatmentWell ShapeNo. per PackProduct Type
446490MultiSorpRound, C Bottom10Frame
446442PolySorpRound, C Bottom5Strip Plate
446469MaxiSorpRound, C Bottom10Strip Plate
446477PolySorpRound10Frame
446639MaxiSorpRound10Microplate Strip
473539PolySorpRound, C Bottom5Microplate Strip
473768MaxiSorpRound, C Bottom5Strip Plate
95029390Universal BindingRound, Flat Bottom (F-well)50Microplate Strip
Catalog number 446490
Price (USD)
432.59
Online exclusive
664.83
Save 232.24
Case of 60
Surface Treatment:
MultiSorp
Well Shape:
Round, C Bottom
No. per Pack:
10
Product Type:
Frame
Price (USD)
432.59
Online exclusive
664.83
Save 232.24
Case of 60

Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.

  • Free-standing modules are easy to handle
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
Binding PropertyVery Hydrophilic
BreakapartBreakable Strips
CertifiedNo
DescriptionHigh optical quality for reliable results
Detection MethodColorimetric
For Use With (Application)For glycans and water soluble proteins
FormatStrip Plate
LidWithout Lid
No. per Case60
No. per Pack10
Packaging10 Pk.
Plate BlockingNone
Product TypeFrame
Target MoleculeGlycans
Volume (Metric) Well350 μL
Volume (Metric) Working250 μL
Array8 x 12
ColorClear
MaterialPolystyrene
No. of Wells96
Surface TreatmentMultiSorp
Well ShapeRound, C Bottom
Unit SizeCase of 60

Frequently asked questions (FAQs)

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with LockWell modules?

Due to the locking feature design of the LockWell modules, the 8-Well Strip Caps for Immuno Standard Modules are not compatible with, and will not seal, the LockWell Modules.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.