Immuno Breakable Modules Clear
Actual product may vary
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Immuno Breakable Modules Clear
Thermo Scientific™

Immuno Breakable Modules Clear

Choose from four surfaces and a variety of wells and configurations with these immuno breakable modules, which offer an optimal choice of products for the specific assay.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface TreatmentWell ShapeNo. per PackProduct Type
446639MaxiSorpRound10Microplate Strip
446442PolySorpRound, C Bottom5Strip Plate
446469MaxiSorpRound, C Bottom10Strip Plate
446477PolySorpRound10Frame
446490MultiSorpRound, C Bottom10Frame
473539PolySorpRound, C Bottom5Microplate Strip
473768MaxiSorpRound, C Bottom5Strip Plate
95029390Universal BindingRound, Flat Bottom (F-well)50Microplate Strip
Catalog number 446639
Price (USD)
-
Surface Treatment:
MaxiSorp
Well Shape:
Round
No. per Pack:
10
Product Type:
Microplate Strip

Choose from four surfaces and a variety of wells and configurations with Thermo Scientific™ Immuno Breakable Modules, which offer reliability, well-to-well reproducibility and an optimal choice of products for the specific assay.

  • Free-standing modules are easy to handle
Specifications
Binding PropertyHydrophilic
BreakapartBreakable Strips
CertifiedYes
DescriptionHigh optical quality for reliable results
Detection MethodColorimetric
For Use With (Application)For antibody sandwich assays
FormatStrip Plate
No. per Case60
No. per Pack10
Packaging10 Pk., 60 Cases
Plate BlockingNone
Product TypeMicroplate Strip
Target MoleculeAntibodies (Glycoproteins)
Volume (Metric) Well320 μL
Volume (Metric) Working250 μL
ColorClear
MaterialPolystyrene
No. of Wells96
Surface TreatmentMaxiSorp
Well ShapeRound
Unit SizeCase of 60

Frequently asked questions (FAQs)

What length of peptide is ideal for binding to the Thermo Scientific MaxiSorp surface? What are the detection limitations?

We have tested and found that a 3 amino acid peptide (Pro, Leu, Gly) cannot be detected when passively adsorbed on the MaxiSorp surface. However, this peptide can be detected when covalently immobilized using CovaLink NH Modules and CovaLink NH2 Modules and Plates. Using covalent immobilization of small peptide residues, you can typically obtain a better orientation of the molecule and reduced problems with antibody recognition of the peptide due to masking of the epitope. We have discovered that a 7 amino acid peptide from the MHC Class II antigen can be detected when adsorbed on the MaxiSorp surface. We state that the detection limitation using the MaxiSorp surface is between 3 and 7 amino acid residues.
One additional note is that detection is contingent upon the orientation of the peptide when immobilized. If the active site is inactivated or hidden at the site facing the solid phase, no detection signal is observed.

Can I bind either single- or double-stranded DNA to the Thermo Scientific MaxiSorp surface?

Single-stranded DNA can be adsorbed to our MaxiSorp surface using approximately 10 µg ssDNA per mL PBS, pH 8.2, although the stability is uncertain. Based on our experience, ssDNA immobilized on the MaxiSorp surface is so loosely bound that it can be removed by stringent washing.
Double-stranded DNA will not bind to the MaxiSorp surface. DNA, however, can be covalently bound to Nunc NucleoLink Strips.

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with LockWell modules?

Due to the locking feature design of the LockWell modules, the 8-Well Strip Caps for Immuno Standard Modules are not compatible with, and will not seal, the LockWell Modules.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.