Are libraries generated with the original Ion Total RNA-Seq Kit (Cat. No. 4466666) compatible with the Ion PGM and Proton systems?
Yes, RNA-Seq libraries prepared using the Ion Total RNA-Seq Kit are compatible with the current Ion PGM and Ion Proton systems.