3-Amino-5-ethyl-1H-pyrazole, 97%
3-Amino-5-ethyl-1H-pyrazole, 97%
3-Amino-5-ethyl-1H-pyrazole, 97%
Thermo Scientific Chemicals

3-Amino-5-ethyl-1H-pyrazole, 97%

CAS: 1904-24-1 | C5H9N3 | 111.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 446710010
Price (TWD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1904-24-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only