Is the MicroSEQ ID Microbial Identification Software v3.0 Bundle for 3500 Instrument (3 licenses) (Cat. No. 4467661) compatible with Windows 10?
No, the only MicroSeq software that is compatible with Windows 10 is MicroSEQ ID Microbial Identification Software, version 3.1.3 (Cat. No. A47132).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.