Product Image
Applied Biosystems™
MicroSEQ™ ID Microbial Identification Software v3.0 (3 licenses)
This easy-to-use software enables you to identify and classify unidentified bacterial or fungal sequences by comparing them to a validatedRead more
Have Questions?
Catalog NumberQuantity
44678641 Kit
Catalog number 4467864
Price (USD)
32,320.00
Each
Quantity:
1 Kit
Price (USD)
32,320.00
Each
This easy-to-use software enables you to identify and classify unidentified bacterial or fungal sequences by comparing them to a validated microbial library.
• A validated library of over 2300 microbial sequences help ensure you get the answers you need
• Automatically identify unknown specimens
• Run plates on Applied Biosystems™ 3500 Series Genetic Analyzers directly via the MicroSEQ™ ID Software
• MicroSEQ™ system is a complete integrated solution, including security, audit, and e-signature features
• Qualification and validation support
Confidently Identify Microbes
The MicroSEQ™ system comprises bacterial libraries of 500 bp sequences and over 2300 validated entries--including extensive coverage of gram-negative nonfermenters, Bacillus, Coryneforms, Mycobacteria, and Staphylococcus. Another library containing more than 1100 entries is available for fungal identification. To help ensure accurate identification of the species of your sequences, we rigorously validate all library entries. And unlike other widely available databases, MicroSEQ™ libraries contain only correctly identified organisms and quality-controlled sequences derived from type strains of preeminent culture collections. The libraries also support sequence polymorphisms, and include bacterial Category A biothreat agents.
Automatically Identify Unknown Specimens
Reviewing each specimen in a project file to make a manual identification call can be tedious and time consuming. The new Auto-ID feature in MicroSEQ™ ID Software v3.0 uses a set of editable parameters to automatically identify unknown specimens. The results table is displayed in the report, and an editable comments field allows additional information or manual identifications to be entered into the same table.
Run Plates on 3500/3500xL Instruments Directly via the MicroSEQ™ ID Software
Applied Biosystems™ 3500 Series Genetic Analyzer users are now able to run plates on these instruments directly via the MicroSEQ™ ID Software v3.0 user interface, without needing Autoanalysis Manager. A single software wizard enables you to set up projects, specimens, and plates; adjust the plate layout via drag and drop; start and monitor the run; and review results.
The MicroSEQ™ System: A Complete, Integrated Solution
Streamlining every step in microbial identification, the Applied Biosystems™ MicroSEQ™ system combines the advantages of MicroSEQ™ ID Software with easy to use PrepMan™ Ultra sample preparation reagents and protocols, MicroSEQ™ bacterial and fungal application kits, and industry-leading thermal cyclers and sequencing systems. To help ensure optimal performance and system-wide integration, we develop and test all components of the system together. The MicroSEQ™ ID Software also includes security, audit, and electronic signature features enabling the software to smoothly integrate into your workflow.
Qualification and Validation Support
To help ensure seamless integration into your laboratory, from installation to routine operation, we offer comprehensive installation and operational qualification (IQ/OQ) and validation support services.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)310 Genetic Analyzer, 3100-Avant Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
LicenseStatic
Operating SystemWindows Vista, Windows 7, Windows XP
Product LineSequaMark
Product TypeID Microbial Identification Software
Quantity1 Kit
SoftwareAnalysis
Unit SizeEach
Frequently asked questions (FAQs)
Which 3500 Data Collection software version is compatible with MicroSEQ ID Microbial Identification Software v3.0?
MicroSEQ ID Microbial Identification Software v3.0 is only compatible with 3500 Data Collection software version 1.1.